Learn More
Niobium(IV) selenide, 99.8% (metals basis), Thermo Scientific Chemicals
Description
It is used as a potential substance for thin film solar cells as back contact material. It is widely used in electricity brush & special bearings because of its conductivity and lubricity property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN3283 |
Solubility Information | Insoluble in water. |
Formula Weight | 250.83 |
Percent Purity | 99.8% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Niobium(IV) selenide |
Safety and Handling
Hazard Category | H301+H331-H373 |
Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
EINECSNumber | 234-811-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.