Alfa Aesar™ Niobium phenoxide

Manufacturer: Alfa Aesar™ 5112004

Quantity 2g

 View more versions of this product

Catalog No. AA5112004

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16576-63-9
    Molecular Formula C30H25NbO5
    Linear Formula Nb(C6H5O)5
    Formula Weight 558.44
    MDL Number MFCD00014076
    Chemical Name or Material Niobium phenoxide

    Specifications

    Formula Weight 558.44
    Physical Form Powder
    CAS 16576-63-9
    Linear Formula Nb(C6H5O)5
    MDL Number MFCD00014076
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Sensitivity Moisture Sensitive
    Melting Point 208°C
    Quantity 2g
    Molecular Formula C30H25NbO5
    Chemical Name or Material Niobium phenoxide

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit