Niobium, plasma standard solution, Nb 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13831AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13831AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula NbCl5 in 2% HF
    MDL Number MFCD00011126
    Chemical Name or Material Niobium

    Specifications

    Quantity 500mL
    DOT Information Hazard Class: 8, Packing Group: II
    Physical Form Liquid
    MDL Number MFCD00011126
    Appearance Liquid
    Molecular Formula NbCl5 in 2% HF
    Health Hazard 1 H300-H310-H330-H314-H318: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Niobium

    H300-H310-H330-H314-H318: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit