Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium, plasma standard solution, Nb 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13831AD

Quantity 50mL

 View more versions of this product

Catalog No. AA13831AD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Liquid
NbCl5 in 2% HF
MFCD00011126
Niobium

Specifications

Liquid
NbCl5 in 2% HF
50mL
Liquid
MFCD00011126
Niobium

Safety and Handling

SDS & Safety Documents

H300-H310-H330-H314-H318: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : Hazard Class: 8, Packing Group: II

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit