Alfa Aesar™ Niobium potassium isopropoxide, 2.5% w/v in isopropanol, 99% (metals basis)

Manufacturer: Alfa Aesar™ 39165AD

Quantity 50mL

 View more versions of this product

Catalog No. AA39165AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21864-23-3
    Linear Formula KNb[OCH(CH3)2]6
    Formula Weight 486.53
    MDL Number MFCD00798516
    UN Number UN1219
    Chemical Name or Material Niobium potassium isopropoxide

    Specifications

    Formula Weight 486.53
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula KNb[OCH(CH3)2]6
    Health Hazard 1 Danger
    Flash Point 12° (54°F)
    Odor Alcohol-like
    Color Pale yellow to brown
    Physical Form Liquid
    CAS 21864-23-3
    Sensitivity Moisture Sensitive
    MDL Number MFCD00798516
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Concentration 2.5% w/v in isopropanol
    Quantity 50mL
    UN Number UN1219
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Chemical Name or Material Niobium potassium isopropoxide
    Assay 99%, (metals basis)

    GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit