Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Niobium rod, 6.35mm (0.25 in.) dia., annealed, 99.8% (metals basis)

Click to view available options
Quantity:
5cm
25cm
100cm
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium,element,niobium-93,powder,foil,rod,vn 1,foil, 0.25mm 0.01in thick,columbio,niobio
23936
CHEBI:33344
niobium
[Nb]
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium,element,niobium-93,powder,foil,rod,vn 1,foil, 0.25mm 0.01in thick,columbio,niobio
23936
CHEBI:33344
niobium
[Nb]
Weight ≈2.7g/cm
Quantity 100cm
Odor Odorless
Physical Form Rod, ≈2.7g/cm,Temper: soft
Percent Purity 99.8%
Chemical Name or Material Niobium rod, 6.35mm (0.25 in.) dia., Annealed, Temper: soft
Assay (metals basis)

EINECSNumber : 231-113-5

TSCA : Yes

Recommended Storage : Ambient temperatures