Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) chloride, 99.8%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
10026-12-7
Cl5Nb
270.16
MFCD00011127
YHBDIEWMOMLKOO-UHFFFAOYSA-I
niobium v chloride,niobium pentachloride,niobium chloride nbcl5,nbcl5,columbium pentachloride,niobium v chloride, puratronic,niobium v chloride trace metals basis,niobium v chloride-nb 20-200ppm ta puratrem
24818
pentachloroniobium
Cl[Nb](Cl)(Cl)(Cl)Cl
10026-12-7
Cl5Nb
270.16
MFCD00011127
YHBDIEWMOMLKOO-UHFFFAOYSA-I
niobium v chloride,niobium pentachloride,niobium chloride nbcl5,nbcl5,columbium pentachloride,niobium v chloride, puratronic,niobium v chloride trace metals basis,niobium v chloride-nb 20-200ppm ta puratrem
24818
pentachloroniobium
Cl[Nb](Cl)(Cl)(Cl)Cl
CAS Min % 99.75
CAS Max % 100.0
Aluminum (Al) 4ppm max.
Assay Percent Range 99.75% min
Linear Formula NbCl5
Fieser 14,214
Merck Index 15, 6643
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in hcl,soluble in ccl4,soluble in ethanol
Bismuth (Bi) 0.5ppm max.
Formula Weight 270.16
Physical Form Fine Crystalline Powder
Percent Purity 99.8%
Boron (B) 2ppm max.
Calcium (Ca) 3ppm max.
Chromium (Cr) 0.5ppm max.
Cobalt (Co) 0.2ppm max.
Copper (Cu) 0.5ppm max.
Iron (Fe) 1ppm max.
Lead (Pb) 1ppm max.
Magnesium (Mg) 1ppm max.
Manganese (Mn) 0.5ppm max.
Nickel (Ni) 0.5ppm max.
Molybdenum (Mo) 3ppm max.
Packaging Glass bottle
Potassium (K) 1ppm max.
Silicon (Si) 3ppm max.
Sodium (Na) 1ppm max.
Solubility Solubility in water: reacts with water. Other solubilities: soluble in hcl, soluble in ccl4, soluble in ethanol
Tantalum (Ta) 25ppm max.
Tin (Sn) 1ppm max.
Titanium (Ti) 3ppm max.
Tungsten (W) 2ppm max.
Vanadium (V) 2ppm max.
Zirconium (Zr) 1ppm max.
Boiling Point 254.0°C
Color Yellow
Melting Point 204.7°C to 209.5°C
Quantity 10g
Chemical Name or Material Niobium(V) chloride, 99.8%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 233-059-8

RTECSNumber : QU0350000

TSCA : TSCA