Learn More
Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Description
It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 248.2°C |
Melting Point | 208.3°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in alcohol,hydrochloric acid,carbon tetrachloride and chloroform. |
Formula Weight | 270.2 |
Odor | Acidic |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335 |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: II |
EINECSNumber | 233-059-8 |
RTECSNumber | QU0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.