Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Niobium(V) ethoxide, 99.999% (metals basis), Ta <500ppm

Preparation of niobium oxide and mixed metal oxides

Manufacturer:  Alfa Aesar™ 1468904

Quantity 2g

 View more versions of this product

Catalog No. AA1468904


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3236-82-6
318.21
UN2924
Niobium(V) ethoxide
Nb(OC2H5)5
MFCD00015122
3678883

Specifications

Ta <500ppm
36° (97°F)
140° to 142°C (0.1mmHg)
2g
Liquid
1.5160
MFCD00015122
3678883
Niobium(V) ethoxide
1.258g/mL
Moisture Sensitive
6°C
3236-82-6
Nb(OC2H5)5
318.21
UN2924
Soluble in organic solvents; Decomposes in water
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit