Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) oxide, 99.95% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
1313-96-8
Nb2O5
265.808
MFCD00011128
ZKATWMILCYLAPD-UHFFFAOYSA-N
niobium oxide,niobium v oxide,niobium pentoxide,diniobium pentaoxide,niobia,niobium pentaoxide,diniobium pentoxide,niobium 5+ oxide,nobum oxde
123105
dioxoniobiooxy(dioxo)niobium
O=[Nb](=O)O[Nb](=O)=O
1313-96-8
Nb2O5
265.808
MFCD00011128
ZKATWMILCYLAPD-UHFFFAOYSA-N
niobium oxide,niobium v oxide,niobium pentoxide,diniobium pentaoxide,niobia,niobium pentaoxide,diniobium pentoxide,niobium 5+ oxide,nobum oxde
123105
dioxoniobiooxy(dioxo)niobium
O=[Nb](=O)O[Nb](=O)=O

For Vacuum Deposition

Assay Percent Range 99.95% (metals basis)
Appearance White 3-12mm Sintered Lump
Solubility Information Soluble in HF. Insoluble in water.
Formula Weight 265.81
Physical Form 3 to 12mm Sintered Lump
Density 4.6
Odor Odorless
Color White
Melting Point 1520°C
Quantity 25g
Chemical Name or Material Niobium(V) oxide
Show More Show Less

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-213-6

TSCA : Yes

Recommended Storage : Ambient temperatures