Alfa Aesar™ Niobium wire, 0.5mm (0.02 in.) dia., 99.96% (metals basis excluding Ta)

Manufacturer: Alfa Aesar™ 14335BY

Quantity 100cm

 View more versions of this product

Catalog No. AA14335BY

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-03-1
    MDL Number MFCD00011126
    Chemical Name or Material Niobium wire, 0.5mm (0.02 in.) dia.

    Specifications

    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD00011126
    CAS 7440-03-1
    Odor Odorless
    Weight ≈1.68g/m
    Quantity 100cm
    Chemical Name or Material Niobium wire, 0.5mm (0.02 in.) dia.
    Assay 99.96%, (metals basis excluding Ta)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit