Specifications
NIOSH | |
Olive/Pink | |
Aluminum | |
To protect against all particulate aerosols, even those with oils present | |
1 Ea. |
40 mm round bottle thread | |
Canister Filter | |
Respirator Canister | |
X-plore™ 6530 |