Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPA Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302941AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Essential component of an SCF-type E3 ligase complex, SCF(NIPA), a complex that controls mitotic entry by mediating ubiquitination and subsequent degradation of cyclin B1 (CCNB1). Its cell-cycle-dependent phosphorylation regulates the assembly of the SCF(NIPA) complex, restricting CCNB1 ubiquitination activity to interphase. Its inactivation results in nuclear accumulation of CCNB1 in interphase and premature mitotic entry. May have an antiapoptotic role in NPM-ALK-mediated signaling events.Specifications
NIPA | |
Polyclonal | |
Unconjugated | |
ZC3HC1 | |
OTTHUMP00000212330; hNIPA; hematopoietic stem/progenitor cell protein 216; nuclear interacting partner of anaplastic lymphoma kinase (ALK); nuclear interaction partner of ALK; nuclear-interacting partner of ALK; nuclear-interacting partner of anaplastic lymphoma kinase; zinc finger C3HC-type protein 1; zinc finger, C3HC type 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51530 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86WB0 | |
ZC3HC1 | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title