Learn More
Cole-Parmer Parker Hannifin Threaded nipple, PFA, 1/4" male NPT Fitting  DFS Item
Manufacturer:  Cole-Parmer 3132100
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Catalog No. 50-632-030