Learn More
Cole-Parmer Parker Hannifin PN-2-P Threaded nipple, PFA, 1/8" male NPT Fitting  DFS Item
Manufacturer:  Cole-Parmer 3132098
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Catalog No. 50-632-029