Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPSNAP3A Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176968MFV450
Description
NIPSNAP3A Polyclonal antibody specifically detects NIPSNAP3A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NIPSNAP3A | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp564D177, FLJ13953, HSPC299, MGC14553, nipsnap homolog 3A (C. elegans), NipSnap3A, NipSnap4, protein NipSnap homolog 3A, Protein NipSnap homolog 4, Target for Salmonella secreted protein C, TassC | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of human NIPSNAP3A. The immunogen is located within amino acids 40 - 90 of NIPSNAP3A. Amino Acid Squence: EFLENFEKNAHLRTAH | |
0.1 mL | |
Neuroscience | |
25934 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction