Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIPSNAP3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559997
Description
Immunogen sequence: VHVLWWNESA DSRAAGRHKS HEDPRVVAAV RESVNYLVSQ QNMLLIP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
The NIPSNAP proteins comprise a family of evolutionarily well-conserved proteins with strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. NIPSNAP2, a novel gene encoding a protein with tyrosine phosphorylation sites and a transmembrane domain, is co-amplified with EGFR in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumors. While NIPSNAP3B is highly expressed skeletal muscle, NIPSNAP3A mRNA levels are low. NIPSNAP3A protein is associated with plasma membrane and partially localized in rafts. NIPSNAP proteins have been suggested to be important in vesicular transport.
Specifications
NIPSNAP3A | |
Polyclonal | |
Unconjugated | |
NIPSNAP3A | |
1700054F22Rik; AI429120; HSPC299; nipsnap homolog 3A; nipsnap homolog 3A (C. elegans); NIPSNAP3A; NipSnap4; Protein NipSnap homolog 3A; protein NipSnap homolog 4; Target for Salmonella secreted protein C; TASSC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UFN0 | |
NIPSNAP3A | |
Recombinant protein corresponding to Human NIPSNAP3A. Recombinant protein control fragment (Product #RP-104340). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction