Learn More
NIST™-Traceable Calibration for Liquid-In-Glass Thermometer
Lists the environmental conditions present at time of calibration
$328.00
Products 1
Catalog Number Mfr. No. CalibrationPoints Price Quantity & Availability  
Catalog Number Mfr. No. CalibrationPoints Price Quantity & Availability  
15-700-327
View Documents
Cole-Parmer
1700603
Each for $328.00
Add to cart
Description
Description
  • Provides traceability to NIST™
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate