Learn More
Beckman Coulter NIST Traceable Latex Particle Size Standards
For calibrating Coulter Counter Aperture Tubes, and verifying their performance.
Manufacturer:  Beckman Coulter 6602798
 View more versions of this product
Catalog No. 15-655-122
Add to cart
Description
Description
  • Use the recommended bead with primary aperture for Aperture Calibration
  • Additional sized beads can be used to verify aperture performance after calibration
  • L10 and L20 standards are included with the Multisizer 4e Coulter Counter Sales Group
  • NIST Traceable
Specifications
Specifications
Particle Size Standard
Calibration and Performance Verification
15 mL
L20
Multisizer 4e and other Coulter Counters
Latex
20 μm