Learn More
Beckman Coulter NIST Traceable Latex Particle Size Standards
For calibrating Coulter Counter Aperture Tubes, and verifying their performance.
$177.00
Specifications
For Use With (Application) Calibration and Performance Verification
Product Type Particle Size Standard
Material Latex
View More Specs
Products 2
Catalog Number Mfr. No. Particle Size Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Price Quantity & Availability  
15-655-123
View Documents Promotion Details
Beckman Coulter
6602796
10 μm
Each for $177.00
Add to cart
15-655-122
View Documents Promotion Details
Beckman Coulter
6602798
20 μm
Each for $177.00
Add to cart
Description
Description
  • Use the recommended bead with primary aperture for Aperture Calibration
  • Additional sized beads can be used to verify aperture performance after calibration
  • L10 and L20 standards are included with the Multisizer 4e Coulter Counter Sales Group
  • NIST Traceable
Specifications
Specifications
Calibration and Performance Verification
Particle Size Standard
Latex