Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nital Etchant 10% Nitric Acid in Methanol, Reagents
Nital Etchant 10% Nitric Acid in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CN104000100C
Quantity | 100 mL |
---|
Specifications
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction