Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nital Etchant 18% w/w Nitric Acid in Methanol, Reagents
Nital Etchant 18% w/w Nitric Acid in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CN2030002.5D1
Quantity | 2.5 L |
---|
Chemical Identifiers
67-56-1, 7697-37-2, 7732-18-5 | |
32.04 | |
Nitric Acid in Methyl Alcohol | |
CO |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
Specifications
Colorless to Light Yellow | |
70°C | |
Nitric Acid in Methyl Alcohol | |
CO | |
32.04 | |
Reagent | |
0.91 | |
2.5 L | |
Liquid |
67-56-1, 7697-37-2, 7732-18-5 | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
18% | |
Glass Bottle | |
<1 | |
Non-sterile | |
Nital Etchant 18% w/w Nitric Acid in Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction