Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.28in), Alloy M

$40.30 - $696.50

Specifications

Chemical Name or Material Nitinol compression coil spring, Alloy M
Name Note Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.)
MDL Number MFCD02091734
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA45951KM View Documents Alfa Aesar™
45951KM
25 pc. Each for $206.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA45951KJ View Documents Alfa Aesar™
45951KJ
5 pc. Each for $40.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA45951KN View Documents Alfa Aesar™
45951KN
100 pc. Each for $696.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Nitinol compression coil spring, Alloy M
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.)
MFCD02091734
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit