Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitinol foil, 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface

Manufacturer:  Alfa Aesar™ 44952FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA44952FF


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD02091734
Nitinol foil, 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface

Specifications

≈0.51g/25x25mm
Odorless
MFCD02091734
25 x 25mm
Af approx. 10°C
Nitinol foil, 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface

Safety and Handling

SDS & Safety Documents

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit