Nitinol Foil, 0.38mm (0.015 in.) thick, Alfa Aesar™

Form: Approx.1.56g/25 x 25mm; Af Approx. 10deg.C; Superelastic; Pickled surface

Manufacturer: Alfa Aesar 44954FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA44954FF

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Foil, 0.38mm (0.015 in.) thick,
    MDL Number MFCD02091734
    Chemical Name or Material Nitinol, Flat annealed

    Specifications

    Quantity 25 x 25mm
    Color Silver-gray
    Physical Form Foil, 0.38mm (0.015 in.) thick,
    Odor Odorless
    MDL Number MFCD02091734
    Appearance Silver-gray foil
    Health Hazard 1 H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
    Health Hazard 2 Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
    Chemical Name or Material Nitinol, Flat annealed

    Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.

    H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit