Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol tension coil spring, Alloy M, Alfa Aesar™

Manufacturer:  Alfa Aesar 45655KM

Quantity 25 pieces

 View more versions of this product

Catalog No. AA45655KM


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-02-0,7440-32-6
Solid
MFCD02091734
Nitinol tension coil spring, Wire diameter 0.75mm, mean coil diameter 5.55mm

Specifications

25 pieces
7440-02-0,7440-32-6
Solid
Nickel: 55.75%; Titanium: 44.25%
Silvery gray
Odorless
MFCD02091734
Nitinol tension coil spring, Wire diameter 0.75mm, mean coil diameter 5.55mm

Safety and Handling

SDS & Safety Documents

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit