Nitinol Wire, 0.25mm (0.01 in.) dia., Alfa Aesar™

Form: Approx. 3.27g/10m; Af Approx. 10deg.C; Superelastic; Oxide surface

Manufacturer: Alfa Aesar 44947H2

Quantity 50m

 View more versions of this product

Catalog No. AA44947H2

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Wire, 0.25mm (0.01 in.) dia.
    MDL Number MFCD02091734
    Chemical Name or Material Nitinol, Alloy N, Straight Annealed

    Specifications

    Quantity 50m
    Color Silver-gray
    Physical Form Wire, 0.25mm (0.01 in.) dia.
    Odor Odorless
    MDL Number MFCD02091734
    Appearance Silver-gray wire
    Health Hazard 1 H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
    Chemical Name or Material Nitinol, Alloy N, Straight Annealed

    H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit