Learn More
Nitinol wire, 0.58mm, Thermo Scientific™
Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface
Specifications
Chemical Name or Material | Nitinol |
---|---|
Color | Silver to Gray |
Name Note | Alloy N, straight annealed |
Appearance | Silver-gray wire |
MDL Number | MFCD02091734 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA44951G6
|
Thermo Scientific Chemicals
44951G6 |
10 m | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
AA44951H2
|
Thermo Scientific Chemicals
44951H2 |
50 m | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
AA44951H5
|
Thermo Scientific Chemicals
44951H5 |
150 m | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nitinol | |
Alloy N, straight annealed | |
MFCD02091734 | |
Wire |
Silver to Gray | |
Silver-gray wire | |
Odorless |
Safety and Handling
H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.