Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol wire, , 0.58mmAlfa Aesar™

Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface

Manufacturer:  Alfa Aesar 44951H2

Quantity 50m

 View more versions of this product

Catalog No. AA44951H2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Wire, 0.58mm (0.023 in.) dia.
MFCD02091734
Nitinol, Alloy N, straight annealed

Specifications

Silver-gray
Silver-gray wire
Wire, 0.58mm (0.023 in.) dia.
Nitinol, Alloy N, straight annealed
50m
Odorless
MFCD02091734

Safety and Handling

SDS & Safety Documents

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit