Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol wire, 1mm, Alfa Aesar™

Form: approx. 49.71g/10m; Af approx. 10°C. Solid in various forms

Manufacturer:  Alfa Aesar 45087H2

Quantity 50m

 View more versions of this product

Catalog No. AA45087H2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Wire, 1.0mm (0.039 in.) dia.
MFCD02091734
Nitinol

Specifications

50m
Silver gray wire
Wire, 1.0mm (0.039 in.) dia.
Nitinol
Silver-gray
Odorless
MFCD02091734

Safety and Handling

SDS & Safety Documents

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit