Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitrate, Ion chromatography standard solution, Specpure™, NO3- 1000μg/mL

Click to view available options
Quantity:
100mL
500mL
MFCD00144920
MFCD00144920
Solubility Information Miscible with water.
Physical Form Liquid
Grade Specpure™
Quantity 100mL
Chemical Name or Material Nitrate Ion chromatography standard solution, NO3- 1000μg/mL

TSCA : Yes

Recommended Storage : Ambient temperatures