Learn More
Anaerobe SystemsSupplier Diversity Partner NITRATE REAGENTS A & B - Nitrate Reagents A (Sulfanilic Acid) and Nitrate Reagent B (1,6 Cleve's Acid), 2 Bottles/Pkg DFS Item
Manufacturer:  Anaerobe SystemsSupplier Diversity Partner AS704
Nitrate Reagents A & B are used for the Nitrate Reduction Test to determine the ability of an organism to reduce nitrate to nitrite or free nitrogen gas. Nitrate Reagent A & B is used to determine if an organism has the ability of reducing nitrate to nitrite or free nitrogen gas. Organisms that have the enzyme nitrate reductase can reduce nitrate into a usable source of nitrogen. Biochemical tests are valuable for identification of anaerobic bacteria. Utilization of nitrate discs, when placed on conventional media, has been described as a rapid method for the detection of nitrate reduction by anaerobic bacteria.
Catalog No. NC0174637