Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrate Standard, 1mL = 1mg NO, Ricca Chemical

Click to view available options
Quantity:
500mL
Packaging:
Poly natural
7757-79-1,7732-18-5,7732-18-5
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
7757-79-1,7732-18-5,7732-18-5
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
CAS Min % 97.0,0.7
CAS Max % 100.0,0.73
Physical Form Liquid
Packaging Poly natural
Equivalence 1 mL = 1 mg NO
Color Colorless
Quantity 500mL
Chemical Name or Material Nitrate Standard