SHOWA™ Nitri-Flex™ Ultimate Gloves

Deliver stretchable, breathable comfort with superior abrasion and cut-resistance

Manufacturer: SHOWA™ 5900S08

 View more versions of this product

Catalog No. 19-120-3529B

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) mechanical and automotive, construction, machine or compartment assembly, recycling, bottling, Glass handling
    Liner Material Kevlar™
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Textured
    Glove Cuff Style Knit
    Material Nitrile
    Size 8
    Coated Area Palm
    Color Yellow, green
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit