Learn More
Honeywell™ Nitri-Knit&trade Nitrile Gloves With Interlocked Liner
High performance supported nitrile gloves offer resistance to cuts, snags, abrasions and punctures.
$71.00 - $72.00
Specifications
Certifications/Compliance 21 CFR 170-199, ASTM D3389-05, 29 CFR 1915.157
Color Blue
Length (English) 12 in.
Length (Metric) 30.5 cm
Grip Design Rough
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-389-B
View Documents
Honeywell™
NK803/8H5
8
Dozen of 12 for $71.00
Case of 12 DZ for $852.00
Add to cart
11-389-C
View Documents
Honeywell™
NK803/9
9
Dozen of 12 for $71.00
Case of 12 DZ for $852.00
Add to cart
11-389-D
View Documents
Honeywell™
NK803/10
10
Dozen of 12 for $71.00
Case of 12 DZ for $852.00
Add to cart
19-160-0135
View Documents
Honeywell™
NK803/11
11
Dozen of 12 for $72.00
Case of 12 DZ for $864.00
Add to cart
Description
Description
  • High nitrile content offers resistance to cuts, snags, abrasions and punctures
  • Ideal for petrochemical refining, degreasing operations, automotive assembly and chemical handling
  • Free of latex proteins which can cause allergic reactions
  • Will not swell, sag or weaken from fat absorption
  • True dip coated glove has a permanent bond of nitrile to inner-liner
Specifications
Specifications
21 CFR 170-199, ASTM D3389-05, 29 CFR 1915.157
12 in.
Rough
Blue
30.5 cm