Learn More
MCR Safety Nitri-Med™ Orange Nitrile, Disposable Gloves, Medical Grade, Powder Free, 4 mil
6010O Nitri-Med, Orange, Medical Grade, Powder Free, 4 mil gloves contain no latex proteins and offer excellent resistance to punctures and tears. Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals.
Specifications
Back Material Nitrile
Certifications/Compliance FDA Compliant
Coating Material No Coating
Color Orange
Exterior Finish Textured Grip
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-812-740
View Documents
MCR Safety
6010OL
Large
Pack of 100 for $16.79
Case of 10 PK for $167.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-741
View Documents
MCR Safety
6010OM
Medium
Pack of 100 for $16.79
Case of 10 PK for $167.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-742
View Documents
MCR Safety
6010OS
Small
Pack of 100 for $16.79
Case of 10 PK for $167.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-743
View Documents
MCR Safety
6010OXL
X-Large
Pack of 100 for $16.79
Case of 10 PK for $167.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-744
View Documents
MCR Safety
6010OXXL
XX-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
  • 4 mil, Orange Powder Free Nitrile
  • Component materials comply with all federal regulations for food contact
  • Medical Grade, Class 1 Medical Device
  • Textured Grip
  • Rolled Cuff
  • 9.5 Inch Length
Specifications
Specifications
Nitrile
No Coating
Textured Grip
Ambidextrous
Nitrile
4 mil
FDA Compliant
Orange
Rolled
Unlined
1 Case (10 boxes of 100)