Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 0.3% (v/v), Ricca Chemical
SDP

Catalog Number p-3251987
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
Color Colorless to Yellow
Concentration 0.3% (v/v)
CAS Min % 98.0, 0.44
CAS Max % 100.0, 0.46
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Nitric Acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit