Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 10.5% (v/v), RS, Ricca Chemical

Click to view available options
Quantity:
1L
Packaging:
Poly natural
7732-18-5
10.5% (v/v)
7732-18-5
10.5% (v/v)
Quantity 1L
Appearance Clear colorless to pale yellow liquid
Synonym dihydrogen oxide,dihydrogen monoxide
Physical Form Liquid
Percent Purity 10.50%
Odor Odorless to slightly pungent
Packaging Poly natural
Solubility Solubility in Water: Infinite

Recommended Storage : Corrosive