Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 50% (v/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 535016SC

Packaging Amber glass safety coated
Quantity 500mL

 View more versions of this product

Catalog No. 535016SC

 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7697-37-2
51.0,51.0
Nitric Acid, 50% (v/v)
49.0,49.0
Liquid

Specifications

Clear colorless to pale-yellow
7732-18-5,7697-37-2
51.0,51.0
Amber glass safety coated
Aqueous Solution
500mL
49.0,49.0
Liquid
50% (v/v)
Nitric Acid, 50% (v/v)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit