Learn More
Nitric Acid, 50% (v/v) Aqueous Solution, Ricca Chemical
Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 535032SC
Packaging Amber Glass Bottle, Safety Cote
Quantity 1 qt.
 View more versions of this product
Catalog No. 535032SC
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Chemical Identifiers
7732-18-5,7697-37-2
51.0,51.0
Nitric Acid, 50% (v/v)
49.0,49.0
Liquid
Specifications
Clear colorless to pale-yellow
7732-18-5,7697-37-2
51.0,51.0
Amber Glass Bottle, Safety Cote
Aqueous Solution
1 qt.
49.0,49.0
Liquid
50% (v/v)
Nitric Acid, 50% (v/v)