Nitric acid, 65-70%, 99.999% (metals basis), Alfa Aesar™

For Use in the Manufacture of Fertilizers and Explosives, in organic Synthesis, in Metallurgy

Manufacturer: Alfa Aesar 87920AB

Quantity 10mL

 View more versions of this product

Catalog No. AA87920AB

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7697-37-2
    Physical Form Liquid
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula HNO3
    Formula Weight 63.01
    MDL Number MFCD00011349
    Chemical Name or Material Nitric acid

    Specifications

    Physical Form Liquid
    Formula Weight 63.01
    Quantity 10mL
    Color Colorless to pale yellow
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 231-714-2
    Vapor Pressure 17mm Hg at 20°C
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula HNO3
    MDL Number MFCD00011349
    CAS 7697-37-2
    Health Hazard 1 H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.
    Odor Pungent
    Chemical Name or Material Nitric acid

    H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit