Learn More
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
Description
Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Boiling Point | 121°C |
Concentration | 65 to 70% |
Density | 1.41 g/mL |
Odor | Pungent |
Quantity | 25 mL |
Physical Form | Liquid |
UN Number | UN2031 |
Merck Index | 14,6577 |
Solubility Information | Miscible with water. |
Show More |
Safety and Handling
Hazard Category | H272-H290-H314-H331 |
Hazard Statement | GHS H Statement H314-H272 Causes severe skin burns and eye damage. May intensify fire oxidizer. |
Precautionary Statement | P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID |
EINECSNumber | 231-714-2 |
RTECSNumber | QU5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.