Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 69.0 to 70.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5L
500mL
Packaging:
Glass Bottle
PVC Coated Glass Bottle

Chemical Identifiers

7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
CAS 7697-37-2
Molecular Formula HNO3
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES [N+](=O)(O)[O-]
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.012
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01
Packaging Glass Bottle
Specific Gravity 1L = 1.41kg
Chemical Name or Material Nitric Acid
Assay 69.0 to 70.0%
Grade Baker Analyzed™ ACS Reagent
Show More Show Less