Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, 69.0% to 70.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Formula Weight 63.01
Percent Purity 69.0 to 70.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Clear Glass Bottle
Specific Gravity 1L = 1.41kg
Chemical Name or Material Nitric Acid