Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Nitric Acid ACS, Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass Bottle
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Color Colorless to Yellow
Quantity 500mL
Linear Formula HNO3
Formula Weight 63.01
Grade ACS
Packaging Glass Bottle
Melting Point -41.06°C
Boiling Point 83.94°C
Chemical Name or Material Nitric Acid