Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals

Catalog Number p-3212996
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Plastic bottle
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
7697-37-2, 7732-18-5
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Melting Point -42.0°C
Boiling Point 122.0°C
CAS Min % 68.0, 30.0
CAS Max % 70.0, 32.0
Density 1.4000g/mL
Heavy Metals (as Pb) 0.2ppm max.
Packaging Glass Bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.4
Quantity 25 mL
Physical Form Liquid Free From Suspended Matter or Sediment
Linear Formula HNO3
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Solubility Information Solubility in water: >100 g/100mL (20°C).
Formula Weight 63.01
Percent Purity 68.0 to 70.0%
Grade ACS Reagent
Chemical Name or Material Nitric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep/Store away from clothing/ combustible materials.
Recommended Storage this material may darken during storage due to

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit