Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, for analysis, ca. 65% solution in water, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
25L
Packaging:
Glass Bottle
Metal Drum
Plastic bottle
7697-37-2,7732-18-5
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2,7732-18-5
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
For analysis
Color Colorless
Quantity 25L
CAS Min % 64.0,34.0
CAS Max % 66.0,36.0
Arsenic (As) 0.01ppm max.
Cadmium (Cd) 0.01ppm max.
Linear Formula HNO3
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Chlorine (Cl) 0.5ppm max.
Density 1.4000g/mL
Copper (Cu) 0.05ppm max.
Formula Weight 63.01
Physical Form Liquid
Percent Purity ≥65%
Grade Analysis
Iron (Fe) 0.2ppm max.
Lead (Pb) 0.05ppm max.
Packaging Metal Drum
Solubility Solubility in water: >100 g/100mL (20°C).
Specific Gravity 1.4
Sulfated Ash 0.0005% max.
Sulfate 1ppm max.
Melting Point -42.0°C
Boiling Point 122.0°C
Chemical Name or Material Nitric acid
Show More Show Less

GHS H Statement
May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep/Store away from clothing/ combustible materials.

Danger