Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
PFA Bottle
Poly Bottle
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2
HNO3
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Quantity 500mL
Formula Weight 63.01
Grade BAKER INSTRA-ANALYZED™ Plus
Packaging PFA Bottle
Specific Gravity 1L = 1.41kg
Chemical Name or Material Nitric Acid