Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid concentrate, For 1L standard solution, 1.0 M HNO3 (1.0N), Honeywell Fluka™

Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

For 1L standard solution, 1.0 M HNO3 (1.0N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Boiling Point 100°C
Density 1.200 g/cm3
Packaging Ampoule
Quantity 6 x 1 Ea.
Physical Form Concentrate
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Chemical Name or Material Nitric acid concentrate, For 1L standard solution, 1.0 M HNO3 (1.0N)

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Nitric acid

Recommended Storage : Room Temp