Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitric acid, Environmental Grade Plus, 68-70%

Manufacturer:  Alfa Aesar™ 38748AU

Quantity 1000mL

 View more versions of this product

Catalog No. AA38748AU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7697-37-2
MFCD00011349
14,6577
Environmental Grade Plus
63.01
UN2031
Nitric acid

Specifications

Colorless to pale yellow
7697-37-2
63.01
UN2031
1.41g/mL
121°C
68 to 70%
1000mL
Liquid, Double Sub-Boiling Quartz Distillation
MFCD00011349
14,6577
Pungent
Nitric acid
Environmental Grade Plus

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID

RTECSNumber : QU5775000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit